Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGACATGATCCTGTAGAACTTC[A/G]GAGGTAAGCCTGGTGAATCAGATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606099 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LGALS8 - galectin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006499.4 | 68 | Intron | NP_006490.3 | |||
NM_201543.2 | 68 | Intron | NP_963837.1 | |||
NM_201544.2 | 68 | Intron | NP_963838.1 | |||
NM_201545.2 | 68 | UTR 5 | NP_963839.1 | |||
XM_011544188.2 | 68 | Intron | XP_011542490.1 | |||
XM_017001274.1 | 68 | Intron | XP_016856763.1 | |||
XM_017001275.1 | 68 | Intron | XP_016856764.1 |
LGALS8-AS1 - LGALS8 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |