Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611849 MIM: 177010 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL44 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRPL44 - mitochondrial ribosomal protein L44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINE2 - serpin family E member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136528.1 | Intron | NP_001130000.1 | ||||
NM_001136530.1 | Intron | NP_001130002.1 | ||||
NM_006216.3 | Intron | NP_006207.1 | ||||
XM_005246641.2 | Intron | XP_005246698.1 | ||||
XM_017004329.1 | Intron | XP_016859818.1 | ||||
XM_017004330.1 | Intron | XP_016859819.1 | ||||
XM_017004331.1 | Intron | XP_016859820.1 | ||||
XM_017004332.1 | Intron | XP_016859821.1 |
Set Membership: |
HapMap |