Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTATTTAACCTCAATTACTTTCT[C/T]ATAGGCTCTATCTTCAAATACAGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147679 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL1RN - interleukin 1 receptor antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000577.4 | Intron | NP_000568.1 | ||||
NM_001318914.1 | Intron | NP_001305843.1 | ||||
NM_173841.2 | Intron | NP_776213.1 | ||||
NM_173842.2 | Intron | NP_776214.1 | ||||
NM_173843.2 | Intron | NP_776215.1 | ||||
XM_005263661.4 | Intron | XP_005263718.1 | ||||
XM_011511121.1 | Intron | XP_011509423.1 |
Set Membership: |
HapMap |