Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATTATGATTGGCATCTTTCAC[A/G]TTTTCATGTGGTACTTTCTATTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MS4A13 - membrane spanning 4-domains A13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012417.2 | 342 | Missense Mutation | ATT,GTT | I,V 7 | NP_001012417.2 | |
NM_001100909.1 | 342 | Missense Mutation | ATT,GTT | I,V 7 | NP_001094379.1 | |
NM_001278320.1 | 342 | Missense Mutation | ATT,GTT | I,V 7 | NP_001265249.1 |
Set Membership: |
HapMap |