Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCCTCCGCAAACCTGCCAGAT[A/C]TCCTTCAAGAAACAACAGCTGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604988 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLCO2B1 - solute carrier organic anion transporter family member 2B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145211.2 | Intron | NP_001138683.1 | ||||
NM_001145212.2 | Intron | NP_001138684.1 | ||||
NM_007256.4 | Intron | NP_009187.1 | ||||
XM_017017157.1 | Intron | XP_016872646.1 |
Set Membership: |
HapMap |