Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGATCCTTCACTAGATACTTTT[C/T]TAGATGCCAGGCCCTCTGAGGTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 172420 | ||||||||||||||||||||
Literature Links: |
PLCG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLCG1 - phospholipase C gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002660.2 | Intron | NP_002651.2 | ||||
NM_182811.1 | Intron | NP_877963.1 | ||||
XM_005260438.2 | Intron | XP_005260495.1 | ||||
XM_011528865.1 | Intron | XP_011527167.1 | ||||
XM_011528867.2 | Intron | XP_011527169.1 |
PLCG1-AS1 - PLCG1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |