Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAATTTCATATGATCCAACCTA[A/C]TATAAAACGGAAAAACGTACACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601242 MIM: 608528 | ||||||||||||||||||||
Literature Links: |
MAP1LC3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP1LC3A - microtubule associated protein 1 light chain 3 alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGU - phosphatidylinositol glycan anchor biosynthesis class U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080476.4 | Intron | NP_536724.1 | ||||
XM_011528542.2 | Intron | XP_011526844.1 | ||||
XM_017027664.1 | Intron | XP_016883153.1 | ||||
XM_017027665.1 | Intron | XP_016883154.1 |