Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAAGAAAAAAAAGAACCTAC[C/T]TCTTGTTGAATTTTGGGTAGTATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614469 MIM: 602933 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRRT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SRRT - serrate, RNA effector molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128852.1 | Intron | NP_001122324.1 | ||||
NM_001128853.1 | Intron | NP_001122325.1 | ||||
NM_001128854.1 | Intron | NP_001122326.1 | ||||
NM_015908.5 | Intron | NP_056992.4 | ||||
XM_005250405.2 | Intron | XP_005250462.1 | ||||
XM_005250406.2 | Intron | XP_005250463.1 | ||||
XM_005250407.2 | Intron | XP_005250464.1 | ||||
XM_005250408.2 | Intron | XP_005250465.1 | ||||
XM_017012290.1 | Intron | XP_016867779.1 | ||||
XM_017012291.1 | Intron | XP_016867780.1 |
TRIP6 - thyroid hormone receptor interactor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |