Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTCTGCGGCTCTTCGCCTTGG[A/T]GCACTTGGAAGATGCATTATCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C14orf37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf37 - chromosome 14 open reading frame 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001872.3 | 12742 | Intron | NP_001001872.2 | |||
NM_001320173.1 | 12742 | Intron | NP_001307102.1 | |||
XM_011536471.2 | 12742 | Intron | XP_011534773.1 | |||
XM_017021029.1 | 12742 | UTR 3 | XP_016876518.1 | |||
XM_017021030.1 | 12742 | UTR 3 | XP_016876519.1 |