Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATGGGCAGCCACACTGTGATGC[C/T]TGTGTGAGGGCCTAAACTTCAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615677 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SERPINA9 - serpin family A member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042518.1 | Intron | NP_001035983.1 | ||||
NM_001284275.1 | Intron | NP_001271204.1 | ||||
NM_001284276.1 | Intron | NP_001271205.1 | ||||
NM_175739.3 | Intron | NP_783866.2 | ||||
XM_011536714.2 | Intron | XP_011535016.1 | ||||
XM_011536715.2 | Intron | XP_011535017.1 | ||||
XM_011536716.2 | Intron | XP_011535018.1 | ||||
XM_011536717.1 | Intron | XP_011535019.1 |
Set Membership: |
HapMap |