Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCTATAAATTTTCTCCTTCCC[A/G]GTGATAAGAGAAAGTGACCTTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C4orf50 - chromosome 4 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011513603.2 | Intron | XP_011511905.1 | ||||
XM_011513604.2 | Intron | XP_011511906.1 | ||||
XM_017008891.1 | Intron | XP_016864380.1 | ||||
XM_017008892.1 | Intron | XP_016864381.1 | ||||
XM_017008893.1 | Intron | XP_016864382.1 | ||||
XM_017008894.1 | Intron | XP_016864383.1 |