Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGACAGCAGGGCCGCAGAACGCG[C/T]TGAGCTGGGCCCGAACGTCTAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611233 MIM: 610369 | ||||||||||||||||||||
Literature Links: |
CDNF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDNF - cerebral dopamine neurotrophic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029954.2 | Intron | NP_001025125.2 | ||||
XM_011519488.2 | Intron | XP_011517790.1 |
HSPA14 - heat shock protein family A (Hsp70) member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278205.1 | Intron | NP_001265134.1 | ||||
NM_016299.3 | Intron | NP_057383.2 |