Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGTCACATAGTGTCAGTGTTAA[A/G]AAAAGGTTGTAGGTTTACAGGTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605988 MIM: 606503 | ||||||||||||||||||||
Literature Links: |
DCLRE1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCLRE1C - DNA cross-link repair 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033855.2 | Intron | NP_001029027.1 | ||||
NM_001033857.2 | Intron | NP_001029029.1 | ||||
NM_001033858.2 | Intron | NP_001029030.1 | ||||
NM_001289076.1 | Intron | NP_001276005.1 | ||||
NM_001289077.1 | Intron | NP_001276006.1 | ||||
NM_001289078.1 | Intron | NP_001276007.1 | ||||
NM_001289079.1 | Intron | NP_001276008.1 | ||||
NM_022487.3 | Intron | NP_071932.2 | ||||
XM_006717491.3 | Intron | XP_006717554.1 | ||||
XM_011519616.1 | Intron | XP_011517918.1 | ||||
XM_011519617.1 | Intron | XP_011517919.1 | ||||
XM_011519619.1 | Intron | XP_011517921.1 | ||||
XM_011519620.2 | Intron | XP_011517922.1 | ||||
XM_011519621.1 | Intron | XP_011517923.1 | ||||
XM_017016556.1 | Intron | XP_016872045.1 | ||||
XM_017016557.1 | Intron | XP_016872046.1 | ||||
XM_017016558.1 | Intron | XP_016872047.1 | ||||
XM_017016559.1 | Intron | XP_016872048.1 | ||||
XM_017016560.1 | Intron | XP_016872049.1 |
SUV39H2 - suppressor of variegation 3-9 homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193424.1 | Intron | NP_001180353.1 | ||||
NM_001193425.1 | Intron | NP_001180354.1 | ||||
NM_001193426.1 | Intron | NP_001180355.1 | ||||
NM_001193427.1 | Intron | NP_001180356.1 | ||||
NM_024670.3 | Intron | NP_078946.1 | ||||
XM_006717503.3 | Intron | XP_006717566.1 | ||||
XM_011519662.2 | Intron | XP_011517964.1 | ||||
XM_017016637.1 | Intron | XP_016872126.1 |