Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAGGGATCAGAGTTTTTAAGAA[C/T]AATTTGGTAAATGGGGAGAAGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600448 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCQ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKCQ - protein kinase C theta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242413.2 | Intron | NP_001229342.1 | ||||
NM_001282644.1 | Intron | NP_001269573.1 | ||||
NM_001282645.1 | Intron | NP_001269574.1 | ||||
NM_001323265.1 | Intron | NP_001310194.1 | ||||
NM_001323266.1 | Intron | NP_001310195.1 | ||||
NM_001323267.1 | Intron | NP_001310196.1 | ||||
NM_006257.4 | Intron | NP_006248.1 | ||||
XM_005252496.4 | Intron | XP_005252553.1 | ||||
XM_005252497.4 | Intron | XP_005252554.1 | ||||
XM_006717465.3 | Intron | XP_006717528.1 | ||||
XM_017016410.1 | Intron | XP_016871899.1 | ||||
XM_017016411.1 | Intron | XP_016871900.1 |