Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCACATTGCTGAGGAGCAGTGA[C/T]TGAAGCTTAATCTCATTGTCTACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR31 - WD repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006615.2 | 1829 | UTR 3 | NP_001006616.1 | |||
NM_001012361.3 | 1829 | UTR 3 | NP_001012361.1 | |||
NM_145241.4 | 1829 | UTR 3 | NP_660284.1 | |||
XM_011518194.2 | 1829 | UTR 3 | XP_011516496.1 |