Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGGGCGCCCCGCTGCTGCCCCC[A/G]CTGCTGCTGCTGCTGCTGTGGGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120215 | ||||||||||||||||||||
Literature Links: |
COL5A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL5A1 - collagen type V alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000093.4 | 449 | Silent Mutation | CCA,CCG | P,P 22 | NP_000084.3 | |
NM_001278074.1 | 449 | Silent Mutation | CCA,CCG | P,P 22 | NP_001265003.1 | |
XM_017014266.1 | 449 | Silent Mutation | CCA,CCG | P,P 22 | XP_016869755.1 |
COL5A1-AS1 - COL5A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |