Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTGTCTGCATTTCTCTCAGAT[C/G]TGCAATCTCTTGGGTCAGTCTGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614178 | ||||||||||||||||||||
Literature Links: |
CRACR2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRACR2A - calcium release activated channel regulator 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144958.1 | Intron | NP_001138430.1 | ||||
NM_032680.3 | Intron | NP_116069.1 | ||||
XM_006719021.3 | Intron | XP_006719084.1 | ||||
XM_011521034.2 | Intron | XP_011519336.1 | ||||
XM_011521036.2 | Intron | XP_011519338.1 | ||||
XM_011521037.2 | Intron | XP_011519339.1 | ||||
XM_011521038.2 | Intron | XP_011519340.1 |