Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCGGTCAGCACCGACTACTGGCT[A/G]TACCTGGAGGAGGGTGTGATTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606405 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CACNG5 - calcium voltage-gated channel auxiliary subunit gamma 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145811.2 | 162 | Silent Mutation | CTA,CTG | L,L 34 | NP_665810.1 |
Set Membership: |
HapMap |