Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTATGGCTTAGGCTTATGCCCTA[C/T]GATCGCCCTCACTGACATCGCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612507 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCA9 - ATP binding cassette subfamily A member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080283.3 | 5160 | UTR 3 | NP_525022.2 | |||
XM_017024011.1 | 5160 | UTR 3 | XP_016879500.1 | |||
XM_017024012.1 | 5160 | Intron | XP_016879501.1 | |||
XM_017024013.1 | 5160 | Intron | XP_016879502.1 | |||
XM_017024014.1 | 5160 | Intron | XP_016879503.1 | |||
XM_017024015.1 | 5160 | Intron | XP_016879504.1 |
Set Membership: |
HapMap |