Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGACAGAATTAAAGCAACTGTA[C/G]GTCTTTACTACCCGCCTGACCAGCT
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 605980 | |||||||||||||||||||||||||||||
Literature Links: |
NOD1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
NOD1 - nucleotide binding oligomerization domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006092.2 | Intron | NP_006083.1 | ||||
XM_005249568.1 | Intron | XP_005249625.1 | ||||
XM_005249572.1 | Intron | XP_005249629.1 | ||||
XM_005249576.1 | Intron | XP_005249633.1 | ||||
XM_006715633.2 | Intron | XP_006715696.1 | ||||
XM_011515079.1 | Intron | XP_011513381.1 | ||||
XM_011515080.2 | Intron | XP_011513382.1 | ||||
XM_011515081.2 | Intron | XP_011513383.1 | ||||
XM_011515083.1 | Intron | XP_011513385.1 | ||||
XM_011515084.1 | Intron | XP_011513386.1 | ||||
XM_011515085.1 | Intron | XP_011513387.1 | ||||
XM_011515087.1 | Intron | XP_011513389.1 | ||||
XM_011515088.2 | Intron | XP_011513390.1 | ||||
XM_017011674.1 | Intron | XP_016867163.1 | ||||
XM_017011675.1 | Intron | XP_016867164.1 |
Set Membership: |
HapMap |