Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGTTCAGCTCCAGACATTTTA[C/G]AAAGGGTAGGAGGAAGGAGGAAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603648 MIM: 610415 | ||||||||||||||||||||
Literature Links: |
COX11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX11 - COX11, cytochrome c oxidase copper chaperone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STXBP4 - syntaxin binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178509.5 | Intron | NP_848604.3 | ||||
XM_005257187.4 | Intron | XP_005257244.1 | ||||
XM_006721797.3 | Intron | XP_006721860.1 | ||||
XM_006721798.3 | Intron | XP_006721861.1 | ||||
XM_017024410.1 | Intron | XP_016879899.1 | ||||
XM_017024411.1 | Intron | XP_016879900.1 | ||||
XM_017024412.1 | Intron | XP_016879901.1 | ||||
XM_017024413.1 | Intron | XP_016879902.1 | ||||
XM_017024414.1 | Intron | XP_016879903.1 | ||||
XM_017024415.1 | Intron | XP_016879904.1 |