Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAGTTTCGGAAGGGCTGTATTA[A/G]CATGTACTATTCCTTCTGCTTGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611825 MIM: 614966 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC46 - leucine rich repeat containing 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033413.3 | Intron | NP_219481.1 | ||||
XM_005257775.4 | Intron | XP_005257832.1 |
MRPL10 - mitochondrial ribosomal protein L10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRN2 - secernin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |