Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGTTCTTGCTTCCTTTTAACTC[C/T]GGAGAAGCCTGTGGGCTTAGAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC93 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC93 - coiled-coil domain containing 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019044.4 | 4405 | UTR 3 | NP_061917.3 | |||
XM_006712600.2 | 4405 | Intron | XP_006712663.1 | |||
XM_011511359.1 | 4405 | UTR 3 | XP_011509661.1 | |||
XM_011511361.1 | 4405 | UTR 3 | XP_011509663.1 | |||
XM_011511362.1 | 4405 | UTR 3 | XP_011509664.1 | |||
XM_011511363.2 | 4405 | Intron | XP_011509665.1 |
LOC101929908 - uncharacterized LOC101929908 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |