Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTCTCTAGGACTGGACTCTTC[C/T]TAAGCAAGTCCGGTATGTAGGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606623 | ||||||||||||||||||||
Literature Links: |
HSD17B6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD17B6 - hydroxysteroid 17-beta dehydrogenase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003725.3 | 314 | UTR 5 | NP_003716.2 | |||
XM_005269207.1 | 314 | UTR 5 | XP_005269264.1 | |||
XM_005269208.1 | 314 | UTR 5 | XP_005269265.1 | |||
XM_005269209.1 | 314 | UTR 5 | XP_005269266.1 | |||
XM_006719672.1 | 314 | UTR 5 | XP_006719735.1 | |||
XM_011538925.1 | 314 | UTR 5 | XP_011537227.1 | |||
XM_011538926.1 | 314 | UTR 5 | XP_011537228.1 | |||
XM_011538927.1 | 314 | UTR 5 | XP_011537229.1 |