Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTGTCAATGTACTGATTAAAA[G/T]GTCTAAAAAAATACTCATAAAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300955 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOOL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOOL - apolipoprotein O like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198450.5 | Intron | NP_940852.3 | ||||
XM_011530859.2 | Intron | XP_011529161.1 | ||||
XM_017029271.1 | Intron | XP_016884760.1 | ||||
XM_017029272.1 | Intron | XP_016884761.1 | ||||
XM_017029273.1 | Intron | XP_016884762.1 |