Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTAACCCAGGAATTTCCCCA[G/T]CACTTCTCAGAGACCCACCAACCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614708 MIM: 194549 | ||||||||||||||||||||
Literature Links: |
SCUBE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCUBE3 - signal peptide, CUB domain and EGF like domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF76 - zinc finger protein 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292032.1 | Intron | NP_001278961.1 | ||||
NM_003427.4 | Intron | NP_003418.2 | ||||
XM_011514846.2 | Intron | XP_011513148.1 | ||||
XM_011514847.2 | Intron | XP_011513149.1 | ||||
XM_011514849.2 | Intron | XP_011513151.1 | ||||
XM_011514850.2 | Intron | XP_011513152.1 | ||||
XM_011514854.2 | Intron | XP_011513156.1 | ||||
XM_011514855.2 | Intron | XP_011513157.1 | ||||
XM_011514856.2 | Intron | XP_011513158.1 | ||||
XM_017011248.1 | Intron | XP_016866737.1 | ||||
XM_017011249.1 | Intron | XP_016866738.1 | ||||
XM_017011250.1 | Intron | XP_016866739.1 | ||||
XM_017011251.1 | Intron | XP_016866740.1 | ||||
XM_017011252.1 | Intron | XP_016866741.1 | ||||
XM_017011253.1 | Intron | XP_016866742.1 | ||||
XM_017011254.1 | Intron | XP_016866743.1 | ||||
XM_017011255.1 | Intron | XP_016866744.1 | ||||
XM_017011256.1 | Intron | XP_016866745.1 | ||||
XM_017011257.1 | Intron | XP_016866746.1 |