Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGGGTCACTCCTGGGTCTGAA[A/G]GCTACCTCTGTTACCATCTGTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607786 MIM: 610569 | ||||||||||||||||||||
Literature Links: |
PCSK9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCSK9 - proprotein convertase subtilisin/kexin type 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP24 - ubiquitin specific peptidase 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015306.2 | Intron | NP_056121.2 | ||||
XM_005270690.3 | Intron | XP_005270747.1 | ||||
XM_017000831.1 | Intron | XP_016856320.1 | ||||
XM_017000832.1 | Intron | XP_016856321.1 | ||||
XM_017000833.1 | Intron | XP_016856322.1 | ||||
XM_017000834.1 | Intron | XP_016856323.1 | ||||
XM_017000835.1 | Intron | XP_016856324.1 | ||||
XM_017000836.1 | Intron | XP_016856325.1 | ||||
XM_017000837.1 | Intron | XP_016856326.1 |