Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATAAGAGAGTGTATTTAAAAAA[A/G]ATAATCAAATGCTTTTTGAAAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607796 MIM: 607867 | ||||||||||||||||||||
Literature Links: |
PHF11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHF11 - PHD finger protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCBTB1 - RCC1 and BTB domain containing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018191.3 | 3709 | UTR 3 | NP_060661.3 | |||
XM_005266441.2 | 3709 | UTR 3 | XP_005266498.1 | |||
XM_011535133.1 | 3709 | UTR 3 | XP_011533435.1 | |||
XM_011535134.1 | 3709 | UTR 3 | XP_011533436.1 | |||
XM_011535135.2 | 3709 | UTR 3 | XP_011533437.1 | |||
XM_011535136.2 | 3709 | Intron | XP_011533438.1 |
SETDB2-PHF11 - SETDB2-PHF11 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |