Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACATTCAGTAAATTTTAAAATGG[G/T]TATGTACTTAAAACTGGGTCAATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603128 | ||||||||||||||||||||
Literature Links: |
SIM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIM1 - single-minded family bHLH transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005068.2 | 5811 | Intron | NP_005059.2 | |||
XM_005267100.2 | 5811 | UTR 3 | XP_005267157.1 | |||
XM_011536072.2 | 5811 | Intron | XP_011534374.1 | |||
XM_017011197.1 | 5811 | UTR 3 | XP_016866686.1 |