Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTTTTGCTTCACTTGGACTATT[C/T]TAAAACACAGATCAACAAACGGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613083 | ||||||||||||||||||||
Literature Links: |
LTN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LTN1 - listerin E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320766.1 | 5790 | UTR 3 | NP_001307695.1 | |||
NM_015565.2 | 5790 | UTR 3 | NP_056380.2 | |||
XM_006723987.3 | 5790 | Intron | XP_006724050.1 | |||
XM_017028316.1 | 5790 | UTR 3 | XP_016883805.1 | |||
XM_017028317.1 | 5790 | UTR 3 | XP_016883806.1 |