Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608396 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9A9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SLC9A9 - solute carrier family 9 member A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173653.3 | 2951 | UTR 3 | NP_775924.1 | |||
XM_011512703.2 | 2951 | Intron | XP_011511005.1 | |||
XM_011512704.2 | 2951 | Intron | XP_011511006.1 | |||
XM_017006202.1 | 2951 | Intron | XP_016861691.1 | |||
XM_017006203.1 | 2951 | Intron | XP_016861692.1 |
Set Membership: |
HapMap |