Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610347 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPRD1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPRD1A - regulation of nuclear pre-mRNA domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303411.1 | 2975 | UTR 3 | NP_001290340.1 | |||
NM_001303412.1 | 2975 | Intron | NP_001290341.1 | |||
NM_001303413.1 | 2975 | Intron | NP_001290342.1 | |||
NM_018170.4 | 2975 | UTR 3 | NP_060640.2 | |||
XM_011526048.1 | 2975 | Intron | XP_011524350.1 | |||
XM_011526049.1 | 2975 | UTR 3 | XP_011524351.1 | |||
XM_011526050.1 | 2975 | UTR 3 | XP_011524352.1 | |||
XM_017025808.1 | 2975 | Intron | XP_016881297.1 |
Set Membership: |
HapMap |