Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613781 MIM: 601955 | ||||||||||||||||||||
Literature Links: |
CCDC125 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC125 - coiled-coil domain containing 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297696.1 | Intron | NP_001284625.1 | ||||
NM_001297697.1 | Intron | NP_001284626.1 | ||||
NM_176816.4 | Intron | NP_789786.2 | ||||
XM_005248458.4 | Intron | XP_005248515.1 | ||||
XM_005248459.4 | Intron | XP_005248516.1 | ||||
XM_005248461.3 | Intron | XP_005248518.1 | ||||
XM_005248463.3 | Intron | XP_005248520.1 | ||||
XM_005248464.3 | Intron | XP_005248521.1 | ||||
XM_006714569.3 | Intron | XP_006714632.1 | ||||
XM_006714570.3 | Intron | XP_006714633.1 | ||||
XM_011543255.2 | Intron | XP_011541557.1 | ||||
XM_011543256.2 | Intron | XP_011541558.1 | ||||
XM_011543257.2 | Intron | XP_011541559.1 | ||||
XM_011543258.2 | Intron | XP_011541560.1 | ||||
XM_011543259.2 | Intron | XP_011541561.1 | ||||
XM_011543260.2 | Intron | XP_011541562.1 | ||||
XM_017009206.1 | Intron | XP_016864695.1 | ||||
XM_017009207.1 | Intron | XP_016864696.1 | ||||
XM_017009208.1 | Intron | XP_016864697.1 | ||||
XM_017009209.1 | Intron | XP_016864698.1 | ||||
XM_017009210.1 | Intron | XP_016864699.1 | ||||
XM_017009211.1 | Intron | XP_016864700.1 |
CDK7 - cyclin dependent kinase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |