Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607725 MIM: 608513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARP2 - poly(ADP-ribose) polymerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042618.1 | 325 | Intron | NP_001036083.1 | |||
NM_005484.3 | 325 | Intron | NP_005475.2 | |||
XM_005267247.3 | 325 | Intron | XP_005267304.1 | |||
XM_017020912.1 | 325 | UTR 5 | XP_016876401.1 |
RPPH1 - ribonuclease P RNA component H1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |