Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609895 MIM: 601791 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASZ1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CASZ1 - castor zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079843.2 | 7436 | UTR 3 | NP_001073312.1 | |||
NM_017766.4 | 7436 | Intron | NP_060236.3 | |||
XM_005263479.3 | 7436 | Intron | XP_005263536.1 | |||
XM_011541635.2 | 7436 | Intron | XP_011539937.1 | |||
XM_017001539.1 | 7436 | UTR 3 | XP_016857028.1 | |||
XM_017001540.1 | 7436 | UTR 3 | XP_016857029.1 | |||
XM_017001541.1 | 7436 | UTR 3 | XP_016857030.1 |
PEX14 - peroxisomal biogenesis factor 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |