Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114019 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDH15 - cadherin 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A31 - solute carrier family 22 member 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242757.1 | 2242 | Missense Mutation | GAG,GGG | E,G 80 | NP_001229686.1 | |
XM_011522897.2 | 2242 | Missense Mutation | GAG,GGG | E,G 80 | XP_011521199.1 | |
XM_011522899.2 | 2242 | Missense Mutation | GAG,GGG | E,G 80 | XP_011521201.1 | |
XM_017022962.1 | 2242 | Missense Mutation | GAG,GGG | E,G 113 | XP_016878451.1 | |
XM_017022963.1 | 2242 | Missense Mutation | GAG,GGG | E,G 87 | XP_016878452.1 | |
XM_017022964.1 | 2242 | Missense Mutation | GAG,GGG | E,G 87 | XP_016878453.1 | |
XM_017022965.1 | 2242 | Missense Mutation | GAG,GGG | E,G 87 | XP_016878454.1 | |
XM_017022966.1 | 2242 | Missense Mutation | GAG,GGG | E,G 87 | XP_016878455.1 |
ZNF778 - zinc finger protein 778 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |