Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603265 MIM: 604161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID3A - AT-rich interaction domain 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005224.2 | 564 | Missense Mutation | AGC,GGC | S,G 37 | NP_005215.1 | |
XM_005259513.4 | 564 | Missense Mutation | AGC,GGC | S,G 37 | XP_005259570.1 | |
XM_005259514.4 | 564 | Missense Mutation | AGC,GGC | S,G 37 | XP_005259571.1 | |
XM_017026445.1 | 564 | Missense Mutation | AGC,GGC | S,G 37 | XP_016881934.1 | |
XM_017026446.1 | 564 | Intron | XP_016881935.1 | |||
XM_017026447.1 | 564 | Intron | XP_016881936.1 |
KISS1R - KISS1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |