Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602296 MIM: 600592 MIM: 602955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4M1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP4M1 - adaptor related protein complex 4 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004722.3 | 2701 | UTR 3 | NP_004713.2 | |||
XM_005250689.4 | 2701 | UTR 3 | XP_005250746.1 | |||
XM_005250690.4 | 2701 | UTR 3 | XP_005250747.1 | |||
XM_006716175.3 | 2701 | UTR 3 | XP_006716238.1 | |||
XM_011516685.2 | 2701 | UTR 3 | XP_011514987.1 | |||
XM_017012790.1 | 2701 | UTR 3 | XP_016868279.1 | |||
XM_017012791.1 | 2701 | UTR 3 | XP_016868280.1 |
MCM7 - minichromosome maintenance complex component 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF6 - TATA-box binding protein associated factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190415.1 | 2701 | UTR 3 | NP_001177344.1 | |||
NM_005641.3 | 2701 | UTR 3 | NP_005632.1 | |||
NM_139315.2 | 2701 | UTR 3 | NP_647476.1 | |||
XM_006716100.2 | 2701 | UTR 3 | XP_006716163.1 | |||
XM_006716101.2 | 2701 | UTR 3 | XP_006716164.1 | |||
XM_006716102.2 | 2701 | UTR 3 | XP_006716165.1 | |||
XM_011516542.2 | 2701 | UTR 3 | XP_011514844.1 | |||
XM_011516543.2 | 2701 | UTR 3 | XP_011514845.1 | |||
XM_017012568.1 | 2701 | UTR 3 | XP_016868057.1 |