Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600305 MIM: 606447 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECI1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ECI1 - enoyl-CoA delta isomerase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNPS1 - RNA binding protein with serine rich domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286625.1 | Intron | NP_001273554.1 | ||||
NM_001286626.1 | Intron | NP_001273555.1 | ||||
NM_001286627.1 | Intron | NP_001273556.1 | ||||
NM_006711.4 | Intron | NP_006702.1 | ||||
NM_080594.3 | Intron | NP_542161.1 | ||||
XM_005255048.1 | Intron | XP_005255105.1 | ||||
XM_005255049.3 | Intron | XP_005255106.1 | ||||
XM_017022874.1 | Intron | XP_016878363.1 |
Set Membership: |
HapMap |