Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133510 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ERCC3 - ERCC excision repair 3, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000122.1 | 2277 | Missense Mutation | CCC,TCC | P,S 735 | NP_000113.1 | |
NM_001303416.1 | 2277 | Missense Mutation | CCC,TCC | P,S 671 | NP_001290345.1 | |
NM_001303418.1 | 2277 | Missense Mutation | CCC,TCC | P,S 671 | NP_001290347.1 | |
XM_011510794.1 | 2277 | Missense Mutation | CCC,TCC | P,S 741 | XP_011509096.1 | |
XM_011510795.1 | 2277 | Missense Mutation | CCC,TCC | P,S 589 | XP_011509097.1 | |
XM_017003583.1 | 2277 | Missense Mutation | CCC,TCC | P,S 583 | XP_016859072.1 |
Set Membership: |
HapMap |