Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613878 MIM: 609499 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
F7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
F7 - coagulation factor VII | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000131.4 | 162 | Silent Mutation | CCA,CCG | P,P 36 | NP_000122.1 | |
NM_001267554.1 | 162 | Intron | NP_001254483.1 | |||
NM_019616.3 | 162 | Intron | NP_062562.1 | |||
XM_006719963.3 | 162 | Intron | XP_006720026.2 | |||
XM_011537474.2 | 162 | Intron | XP_011535776.2 | |||
XM_011537475.2 | 162 | Intron | XP_011535777.2 | |||
XM_011537476.2 | 162 | Intron | XP_011535778.1 |
MCF2L - MCF.2 cell line derived transforming sequence like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |