Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606945 | ||||||||||||||||||||
Literature Links: |
LDLR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDLR - low density lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000527.4 | Intron | NP_000518.1 | ||||
NM_001195798.1 | Intron | NP_001182727.1 | ||||
NM_001195799.1 | Intron | NP_001182728.1 | ||||
NM_001195800.1 | Intron | NP_001182729.1 | ||||
NM_001195803.1 | Intron | NP_001182732.1 | ||||
XM_011528010.1 | Intron | XP_011526312.1 |