Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612309 MIM: 173610 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
F5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
F5 - coagulation factor V | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SELP - selectin P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003005.3 | Intron | NP_002996.2 | ||||
XM_005245435.1 | Intron | XP_005245492.1 | ||||
XM_005245436.3 | Intron | XP_005245493.1 | ||||
XM_005245438.1 | Intron | XP_005245495.1 | ||||
XM_005245439.1 | Intron | XP_005245496.1 | ||||
XM_005245440.1 | Intron | XP_005245497.1 |