Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTCCACCTCAACCAGGAGGAC[A/G]AAAGCACCATACTGTGGGAAGGGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608861 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1H PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP6V1H - ATPase H+ transporting V1 subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015941.3 | Intron | NP_057025.2 | ||||
NM_213619.2 | Intron | NP_998784.1 | ||||
NM_213620.2 | Intron | NP_998785.1 | ||||
XM_006716455.2 | Intron | XP_006716518.1 | ||||
XM_011517542.1 | Intron | XP_011515844.1 |
Set Membership: |
HapMap |