Search Thermo Fisher Scientific
CTCTACAAATCGTCAACACTCAACA[A/G]ATGCTAATTTTTTTCCTCTCTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613543 MIM: 610012 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO5A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLCO5A1 - solute carrier organic anion transporter family member 5A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146008.1 | 6032 | Intron | NP_001139480.1 | |||
NM_001146009.1 | 6032 | Intron | NP_001139481.1 | |||
NM_030958.2 | 6032 | Intron | NP_112220.2 | |||
XM_005251313.2 | 6032 | UTR 3 | XP_005251370.1 | |||
XM_017013883.1 | 6032 | UTR 3 | XP_016869372.1 | |||
XM_017013884.1 | 6032 | Intron | XP_016869373.1 | |||
XM_017013885.1 | 6032 | UTR 3 | XP_016869374.1 | |||
XM_017013886.1 | 6032 | Intron | XP_016869375.1 |
SULF1 - sulfatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |