Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN8 - calpain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143962.1 | 1579 | Missense Mutation | AAA,GAA | K,E 684 | NP_001137434.1 | |
XM_017001265.1 | 1579 | Missense Mutation | AAA,GAA | K,E 523 | XP_016856754.1 | |
XM_017001266.1 | 1579 | Missense Mutation | AAA,GAA | K,E 457 | XP_016856755.1 | |
XM_017001267.1 | 1579 | Intron | XP_016856756.1 | |||
XM_017001268.1 | 1579 | Intron | XP_016856757.1 |