Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAATCTTCATGCCTGGTCATGA[C/T]TACAATTGGTCATTTTATTTAACTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171885 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTFR1 - mitochondrial fission regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145838.1 | Intron | NP_001139310.1 | ||||
NM_001145839.1 | Intron | NP_001139311.1 | ||||
NM_014637.3 | Intron | NP_055452.3 | ||||
XM_006716484.2 | Intron | XP_006716547.2 | ||||
XM_011517626.1 | Intron | XP_011515928.1 | ||||
XM_011517627.2 | Intron | XP_011515929.1 | ||||
XM_011517628.2 | Intron | XP_011515930.1 |
PDE7A - phosphodiesterase 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242318.2 | Intron | NP_001229247.1 | ||||
NM_002603.3 | Intron | NP_002594.1 | ||||
XM_011517540.2 | Intron | XP_011515842.1 | ||||
XM_017013538.1 | Intron | XP_016869027.1 |
Set Membership: |
HapMap |