Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAATTAATGAGTAAAGGAGGCA[C/T]GGCAGCAATAATCTGTTCAGATATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300572 MIM: 300798 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADGRG2 - adhesion G protein-coupled receptor G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079858.2 | Intron | NP_001073327.1 | ||||
NM_001079859.2 | Intron | NP_001073328.1 | ||||
NM_001079860.2 | Intron | NP_001073329.1 | ||||
NM_001184833.1 | Intron | NP_001171762.1 | ||||
NM_001184834.1 | Intron | NP_001171763.1 | ||||
NM_001184835.1 | Intron | NP_001171764.1 | ||||
NM_001184836.1 | Intron | NP_001171765.1 | ||||
NM_001184837.1 | Intron | NP_001171766.1 | ||||
NM_005756.3 | Intron | NP_005747.2 | ||||
XM_006724455.3 | Intron | XP_006724518.1 | ||||
XM_011545434.1 | Intron | XP_011543736.1 | ||||
XM_011545435.2 | Intron | XP_011543737.1 |
LOC101928415 - uncharacterized LOC101928415 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHKA2 - phosphorylase kinase regulatory subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |