Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTCACACACAAACACATGGTGC[A/G]TCTCCAGAGGCCATGGAAGTGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604110 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRG1 - adhesion G protein-coupled receptor G1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ADGRG3 - adhesion G protein-coupled receptor G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308360.1 | Intron | NP_001295289.1 | ||||
NM_170776.4 | Intron | NP_740746.4 | ||||
XM_005255842.3 | Intron | XP_005255899.1 | ||||
XM_006721170.3 | Intron | XP_006721233.1 | ||||
XM_011522951.2 | Intron | XP_011521253.1 | ||||
XM_011522953.2 | Intron | XP_011521255.1 | ||||
XM_011522954.1 | Intron | XP_011521256.1 |
Set Membership: |
HapMap |