Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTTGACTGGTGTGAGTTGGTTG[C/T]GGTTAAAATGTAAACAGTCAGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTFR1 - mitochondrial fission regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145838.1 | Intron | NP_001139310.1 | ||||
NM_001145839.1 | Intron | NP_001139311.1 | ||||
NM_014637.3 | Intron | NP_055452.3 | ||||
XM_006716484.2 | Intron | XP_006716547.2 | ||||
XM_011517626.1 | Intron | XP_011515928.1 | ||||
XM_011517627.2 | Intron | XP_011515929.1 | ||||
XM_011517628.2 | Intron | XP_011515930.1 |